missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ YC 1

Description
Nitric oxide-independent activator of soluble guanylyl cyclase (sGC). Significantly elevates cGMP levels and inhibits collagen-stimulated aggregation of washed rabbit platelets (IC50 = 14.6 μM); induces relaxation in denuded phenylephrine-contracted rabbit aortic rings (EC50 = 1.9 μM). Also displays antiproliferative activity in vitro and in vivo by inducing G1 cell cycle arrest in two human hepatocellular carcinoma (HCC) cell lines, and in HCC xenografts in athymic SCID mice. Exhibits low cytotoxicity in non-malignant cells.
Specifications
Specifications
| Kemisk navn eller materiale | YC 1 |
| Mængde | 10 mg |
| Formel vægt | 304.34 |
| Renhed | >99% |
| Molekylvægt (g/mol) | 304.35 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?