Få mere at vide
Yangonin, TRC
Artikelnummer. 30135503
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30135503 25 mg -
30004133 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135503 Leverandør Toronto Research Chemicals Leverandørnr. TRCY10055025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 500-62-9
Molekylær formel C15 H14 O4
Molekylvægt (g/mol) 258.27
Synonym 4-Methoxy-6-[(1E)-2-(4-methoxyphenyl)ethenyl]-2H-pyran-2-one, 4-Methoxy-6-[β-(p-anisyl)vinyl]-α-pyrone, 6-(p-Methoxystyryl)-4-methoxy-α-pyrone;
IUPAC navn 4-methoxy-6-[(E)-2-(4-methoxyphenyl)ethenyl]pyran-2-one
SMIL COC1=CC(=O)OC(=C1)\C=C\c2ccc(OC)cc2
Tekniske data
Kemisk navn eller materiale Yangonin
Mængde 25 mg
InChI formel InChI=1S/C15H14O4/c1-17-12-6-3-11(4-7-12)5-8-13-9-14(18-2)10-15(16)19-13/h3-10H,1-2H3/b8-5+
Formel vægt 258.0892