Få mere at vide
Xylan, TRC
Artikelnummer. 17997254
Skift visning
Mængde:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17997254 10 g
17928584 25 g
17927704 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17997254 Leverandør Toronto Research Chemicals Leverandørnr. TRCX74801010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 9014-63-5
Molekylær formel C5H10O6
Molekylvægt (g/mol) 166.13
Synonym Xylans, CMT 85, D-Xylan, DP 42
IUPAC navn (2R,3R,5S,6S)-tetrahydropyran-2,3,4,5,6-pentol
SMIL O[C@@H]1O[C@H](O)[C@@H](O)C(O)[C@H]1O
Tekniske data
Kemisk navn eller materiale Xylan
Mængde 10 g
InChI formel InChI=1S/C5H10O6/c6-1-2(7)4(9)11-5(10)3(1)8/h1-10H/t1?,2-,3+,4-,5+
Formel vægt 166.0477