missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
XRCC6BP1 Mouse anti-Human, Clone: OTI5F3, liquid, TrueMAB™
Beskrivelse
The protein encoded by this gene is amplified in glioblastomas and interacts with the DNA binding subunit of DNA-dependent protein kinase. This kinase is involved in double-strand break repair (DSB), and higher expression of the encoded protein increases the efficiency of DSB. In addition, comparison to orthologous proteins strongly suggests that this protein is a metalloprotease important in the biosynthesis of mitochondrial ATPase. Several transcript variants encoding different isoforms have been found for this gene.
Tekniske data
Tekniske data
| Antigen | XRCC6BP1 |
| Applikationer | Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Monoclonal |
| Klon | OTI5F3 |
| Koncentration | 1 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | ATP23 |
| Genadgangsnr. | Q9Y6H3 |
| Gene Alias | KUB3, XRCC6BP1 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?