missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ XRCC2 Monoclonal Antibody (XRCC2 7B7/3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA124927
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MA1-24927 detects XRCC2 from human samples.
The x-ray repair cross-complementing (XRCC) proteins are responsible for efficiently repairing and maintaining genetic stability following DNA base damage. These genes share sequence similarity with the yeast DNA repair protein Rad5. XRCC2 and XRCC3 are both involved in maintaining chromosome stability during cell division. XRCC2 is required for efficient repair of DNA double-strand breaks by homologous recombination between sister chromatids, and XRCC3 interacts directly with Rad51 to cooperate with Rad51 during recombinational repair.
Tekniske data
| XRCC2 | |
| Monoclonal | |
| 1.49 mg/mL | |
| PBS with no preservative | |
| O43543 | |
| XRCC2 | |
| Recombinant full length protein (Human). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| XRCC2 7B7/3 | |
| Unconjugated | |
| XRCC2 | |
| 4921524O04Rik; 8030409M04Rik; DKFZp781P0919; DNA repair protein XRCC2; FANCU; RAD51; RecA; recA/RAD51 family protein; RGD1564823; X-ray repair complementing defective repair in Chinese hamster cells 2; X-ray repair cross complementing 2; X-ray repair cross complementing protein 2; X-ray repair cross-complementing protein 2; Xrcc2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 7516 | |
| 4°C, do not freeze | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion