missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
XK X-linked Kx blood group Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05963FR
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
XK X-linked Kx blood group Polyclonal specifically detects XK X-linked Kx blood group in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Tekniske data
| XK X-linked Kx blood group | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Kell blood group precursor (McLeod phenotype), Kell complex 37 kDa component, KX, Kx antigen, membrane transport protein XK, X1k, XK, Kell blood group complex subunit (McLeod syndrome), XKR1XK-related protein 1, X-linked Kx blood group (McLeod syndrome), XRG1 | |
| Produced in rabbits immunized with E. coli-derived Human XK X-linked Kx blood group fragment. | |
| 0.1 mL | |
| Neuroscience | |
| 7504 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion