Få mere at vide
Xipamide, LoGiCal
Artikelnummer. 30459777
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459777 10 mg
1 options
Artikelnummer. 30459777 Leverandør LoGiCal Leverandørnr. LGCFOR1214.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Xipamide
CAS 14293-44-8
Analyt- eller komponentnavne Xipamide
Forsendelsestilstand Room Temperature
Molekylær formel C15 H15 Cl N2 O4 S
Certificeringer/compliance ISO 9001
Synonym Benzamide, 5-(aminosulfonyl)-4-chloro-N-(2,6-dimethylphenyl)-2-hydroxy-, 2',6'-Salicyloxylidide, 4-chloro-5-sulfamoyl- (8CI), Aquaphor, Aquaphor (diuretic), BE 1293, BEI 1293, Chronexan, Diurex, Diurex (Lacer), Diurexan, Lumitens, MJF 10938, Xipamide
InChI formel InChI=1S/C15H15ClN2O4S/c1-8-4-3-5-9(2)14(8)18-15(20)10-6-13(23(17,21)22)11(16)7-12(10)19/h3-7,19H,1-2H3,(H,18,20)(H2,17,21,22)
Løsningstype Neat
SMIL Cc1cccc(C)c1NC(=O)c2cc(c(Cl)cc2O)S(=O)(=O)N
IUPAC navn 4-chloro-N-(2,6-dimethylphenyl)-2-hydroxy-5-sulfamoylbenzamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 354.81
Mængde 10 mg
Formel vægt 354.0441 g/mol