Få mere at vide
Xipamide-d6, TRC
Artikelnummer. 30454598
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30454598 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454598 Leverandør Toronto Research Chemicals Leverandørnr. TRCX7451521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Xipamide-d6
CAS 1330262-09-3
Molekylær formel C15H9D6ClN2O4S
Mængde 1 mg
Synonym 5-(Aminosulfonyl)-4-chloro-N-(2,6-dimethylphenyl)-2-hydroxy-benzamide-d6, 4-Chloro-5-sulfamoyl-2',6'-salicyloxylidide-d6, Aquaphor-d6, Chronexan-d6, Diurex-d6, Lumitens-d6
InChI formel InChI=1S/C15H15ClN2O4S/c1-8-4-3-5-9(2)14(8)18-15(20)10-6-13(23(17,21)22)11(16)7-12(10)19/h3-7,19H,1-2H3,(H,18,20)(H2,17,21,22)/i1D3,2D3
SMIL [2H]C([2H])([2H])c1cccc(c1NC(=O)c2cc(c(Cl)cc2O)S(=O)(=O)N)C([2H])([2H])[2H]
Molekylvægt (g/mol) 360.85
Formel vægt 360.0818