Learn More
Trajan Scientific and Medical™ XCHANGE™ CTC PAL-XT Syringe for LC
Product Code. 15279273
Change view
Click to view available options
Volume (Metric):
100 μL
250 μL
500 μL
1 mL
Diameter (Metric) Inner:
0.17 mm
0.375 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Diameter (Metric) Inner Unit Size
15279273 1 mL 0.375 mm Each
15259273 500 μL 0.375 mm Each
15209273 100 μL 0.17 mm Each
15239273 250 μL 0.375 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15279273 Supplier Trajan Scientific and Medical™ Supplier No. 2928830
Forventet forsendelse: 23-12-2026
The knowledge and expertise that goes into manufacturing an SGE syringe gives outstanding performance and superior quality
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
Specifications
Description CTC PAL-XT Syringe for LC
Needle Gauge 22 G
Diameter (Metric) Inner Needle 0.375 mm
Diameter (Metric) Inner 0.375 mm
Diameter (Metric) Outer 0.71 mm
Diameter (Metric) Outer Needle 0.71 mm
For Use With (Application) For liquid handling and LC
Product Type LC Tip
Volume (Metric) 1 mL
Needle Point Style LC