Learn More
Honeywell Safety Products™ XC Replacement Lens, Clear
Description
- For use in any Pulsafe XC frame.
- General use and protection against impact (45m/s) during mechanical work including grinding, turning and milling.
- 9 base-curved high coverage lens.
- Total wraparound coverage for exceptional peripheral vision and complete protection.
Specifications
Certifications/Compliance | EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1144, EC Attestation Number EC1144 |
Lens Coating | Scratch-resistant |
Product Type | Safety glasses |
For Use With (Equipment) | Any Pulsafe XC frame |
For Use With (Application) | Administration, General use and protection against impact including grinding, turning, milling. |
No. per Pack | 10/Pk. |
Lens Material | Polycarbonate |