Learn More
Honeywell Safety Products™ XC Replacement Lens, Clear
Catalog Number p-8492003
Easily fitting into any Pulsafe XC frame, this clear replacement lens provides full, wraparound coverage and protection. Honeywell Safety Products™ XC Replacement Lens is suitable for general use and protection against impact and comes with a scratch-resistant lens.
  • For use in any Pulsafe XC frame.
  • General use and protection against impact (45m/s) during mechanical work including grinding, turning and milling.
  • 9 base-curved high coverage lens.
  • Total wraparound coverage for exceptional peripheral vision and complete protection.
TRUSTED_SUSTAINABILITY
Certifications/Compliance EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1144, EC Attestation Number EC1144
Lens Coating Scratch-resistant
Product Type Safety glasses
For Use With (Equipment) Any Pulsafe XC frame
For Use With (Application) Administration, General use and protection against impact including grinding, turning, milling.
No. per Pack 10/Pk.
Lens Material Polycarbonate