Få mere at vide
Xanthurenic Acid-d4, TRC
Artikelnummer. 30449809
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30449809 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449809 Leverandør Toronto Research Chemicals Leverandørnr. TRCX74350225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Xanthurenic acid 3,5,6,7-d4
CAS 1329611-28-0
Renhedsgrad noter HPLC
Molekylær formel C10 2H4 H3 N O4
Mængde 25 mg
Synonym 3,5,6,7-Tetradeuterio-4,8-dihydroxyquinoline-2-carboxylic acid, Xanthurenic acid-d4, Xanthurenic acid 3,5,6,7-D4, 4,8-Dihydroxy-2-quinolinecarboxylic-3,5,6,7-d4 acid, 4,8-Dihydroxy-3,5,6,7-d4-quinaldic acid, 8-Hydroxykynurenic acid-d4
InChI formel InChI=1S/C10H7NO4/c12-7-3-1-2-5-8(13)4-6(10(14)15)11-9(5)7/h1-4,12H,(H,11,13)(H,14,15)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(O)c2nc(C(=O)O)c([2H])c(O)c2c1[2H]
IUPAC navn 3,5,6,7-tetradeuterio-4,8-dihydroxyquinoline-2-carboxylic acid
Molekylvægt (g/mol) 209.19
Formel vægt 209.0626
Procent renhed >95