missing translation for 'onlineSavingsMsg'
Få mere at vide

Xanthinol Nicotinate, TRC

Artikelnummer. 30454213
Change view
Click to view available options
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30454213 1 mg
30451395 10 mg
30453233 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30454213 Supplier Toronto Research Chemicals Supplier No. TRCX4999601MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 437-74-1
Molekylær formel C13 H21 N5 O4 . C6 H5 N O2
Molekylvægt (g/mol) 434.45
Synonym 3-Pyridinecarboxylic acid, compd. with 3,7-dihydro-7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-1,3-dimethyl-1H-purine-2,6-dione (1:1) (9CI, ACI), 1H-Purine-2,6-dione, 3,7-dihydro-7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-1,3-dimethyl-, mono-3-pyridinecarboxylate (salt) (9CI), Nicotinic acid, compd. with 7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]theophylline (1:1) (8CI), Theophylline, 7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-, mononicotinate (salt) (8CI), Theophylline, 7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-, nicotinate (6CI), 7-[2-Hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]theophylline nicotinate, 7-[2-Hydroxy-3-[N-(2-hydroxyethyl)-N-methylamino]propyl]-1,3-dimethylxanthine pyridine-3-carboxylate, 7-[2-Hydroxy-3-[N-methyl-N-(2-hydroxyethyl)amino]-propyl]-theophylline nicotinate, 7-[3-(Methyl-2-hydroxyethylamino)-2-hydroxypropyl)]theophylline nicotinate, Angioamin, Angiomin, Complamex, Complamin, Complamin retard, Contamex, Landrina 300, NSC 113217, Sadamine, SK 331A, Stenalgil, Teonicol, Vedrin, Xanthinol niacinate, Xanthinol nicotinate, Xantinol nicotinate, Xavin
IUPAC navn 7-[2-hydroxy-3-[2-hydroxyethyl(methyl)amino]propyl]-1,3-dimethylpurine-2,6-dione;pyridine-3-carboxylic acid
SMIL CN(CCO)CC(O)Cn1cnc2N(C)C(=O)N(C)C(=O)c12.OC(=O)c3cccnc3

Specifications

Kemisk navn eller materiale Xanthinol Nicotinate
Mængde 1 mg
InChI formel InChI=1S/C13H21N5O4.C6H5NO2/c1-15(4-5-19)6-9(20)7-18-8-14-11-10(18)12(21)17(3)13(22)16(11)2;8-6(9)5-2-1-3-7-4-5/h8-9,19-20H,4-7H2,1-3H3;1-4H,(H,8,9)
Formel vægt 434.1914
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.