missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human XAGE2 The XAGE2 Recombinant Protein Antigen is derived from E. coli. The XAGE2 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Protein længde | PTPDQKREDDQGAAEIQVPDLEADLQ |
| Renhed | >80% by SDS-PAGE and Coomassie blue staining |
| Fælles navn | XAGE2 Recombinant Protein Antigen |
| Indhold og opbevaring | Store at −20°C short term. Avoid freeze-thaw cycles. |
| Formulering | PBS and 1M Urea, pH 7.4 |
| Til brug med (applikation) | AC |
| Gene Alias | CT12.2GAGED3, G antigen family D member 3 |
| Gen symbol | XAGE2 |
| Etikettype | Unlabeled |
| Molekylvægt (g/mol) | 21 kDa |
| Show More |
For Research Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?