Få mere at vide
Tocris Bioscience™ XAC
GREENER_CHOICE
Artikelnummer. 18034972
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18034972 10 mg
18015422 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18034972 Leverandør Tocris Bioscience™ Leverandørnr. 3200/10
Adenosine receptor antagonist
Adenosine receptor antagonist (IC50 values are 1.8 and 114 nM at A1 and A2 receptors respectively). Attenuates adenosine-induced vasodilation and exhibits proconvulsant activity in vivo.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1962928-23-9
Molekylær formel C21H30Cl2N6O4
Tekniske data
Kemisk navn eller materiale XAC
Mængde 10 mg
Formel vægt 464.95
Renhed >98%
Molekylvægt (g/mol) 501.41