Få mere at vide
Arcis Sample Prep Kit
Inkluderer:
Lyse og buffer
Pakningsstørrelse:
Pakke med 50
Beskrivelse
- Rapid DNA/RNA extraction (less than 3 minutes)
- Suitable for 50 reactions
- No instrumentation required
- Simple, stable process without heating, cooling, spin, or wash steps
- Two-step, two-tube protocol (lysis and buffer)
- DNA stable for 2 weeks
- Extract is suitable for use in PCR and qPCR reactions using standard and fast cycling protocols
- Shipped and stored at ambient temperature
Tekniske data
Tekniske data
| Type | DNA/RNA Extraction Kit |
| Holder | Two-tube System (Lysis and Buffer) |
| Inkluderer | Lyse og buffer |