Learn More
Epredia X1000 Round Coverslip dia. 13mm #1 (0.13-0.16mm)
Brand: Epredia CB00130RA120MNZ0
362.98 DKK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
X1000 Round Coverslip dia. 13mm #1 (0.13-0.16mm)
This item is not returnable.
View return policy
Product Title
Epredia X1000 Round Coverslip dia. 13mm #1 (0.13-0.16mm) >
Spot an opportunity for improvement?Share a Content Correction