missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Violet Red Bile Glucose Agar (ISO)
missing translation for 'orderingAttributeHoverText'
Mængde:
10 Plates
missing translation for 'unitSize'
Pakke med 10
Description
Enhance the detection of glucose-fermenting bacteria with Violet Red Bile Glucose Agar (ISO), designed for selective isolation.
Specifications
Specifications
| Farve | Transparent |
| Indhold og opbevaring | 2°C to 12°C |
| Format | 90 mm |
| Fremstillingsoprindelse | Germany |
| Produkttype | Agar Plate |
| Fyld volumen | 17 mL |
| Mængde | 10 Plates |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction