missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WWP2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00072AF488
Additional Details : Weight : 0.00970kg
Description
WWP2 Polyclonal specifically detects WWP2 in Human samples. It is validated for ELISA.Specifications
WWP2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
11060 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 488 | |
AIP2Nedd-4-like ubiquitin-protein ligase, atrophin-1 interacting protein 2, Atrophin-1-interacting protein 2, EC 6.3.2, EC 6.3.2.-, NEDD4-like E3 ubiquitin-protein ligase WWP2, WW domain containing E3 ubiquitin protein ligase 2, WW domain-containing protein 2, WWp2-like | |
Produced in rabbits immunized with purified, recombinant WWP2 (O00308; Met1-Glu870). | |
0.1 mL | |
Primary | |
Human | |
Purified |