Learn More
Micronova™ WrapMop™ Extension Handles and Adapters
Aluminum extension handles and adapters for Micronova Wrapmops
Brand:  Micronova™ ALUMEH-18
 View more versions of this product
Product Code. 12791635
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • For use with Micronova WrapMop series
  • NovaPoly WrapMop model numbers WM612-14 and WM612-18
  • MegaTex WrapMop model numbers WM62012-14 and WM62012-18
  • Microfiber WrapMop model numbers WM6AA12-14 and WM6AA12-18
Specifications
Specifications
Extension Handle
72 to 216 in.
Aluminum
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number036095-000 Certificate TypeCertificate of Compliance Date12/01/2023 Catalog NumberALUMEH-18
Lot Number033496-000 Certificate TypeCertificate of Compliance Date28/07/2022 Catalog NumberALUMEH-18
Lot Number031026-000 Certificate TypeCertificate of Compliance Date07/07/2022 Catalog NumberALUMEH-18
Lot Number026514-000 Certificate TypeCertificate of Compliance Date06/07/2021 Catalog NumberALUMEH-18
Lot Number025702-000 Certificate TypeCertificate of Compliance Date17/05/2021 Catalog NumberALUMEH-18
Promotions
Promotions
Product Content Correction
Product Title
Micronova™ WrapMop™ Extension Handles and Adapters > Aluminum extension handle: 72 to 216in. (182.9 to 548.6cm)