Få mere at vide
Techsit Worktech Laboratory ESD Low Chair
Beskrivelse
- ESD Cleanroom vinyl low chair on ESD Castors
- Black ESD/Anti-static Vinyl that is easy to clean
- Fully adjustable Seat height, back height, and back rake
- Ergonomically designed and contoured seat and back
- Cleanroom class 7 suitable
- BS EN61340-5-1
- Conductivity 104 - 106 Ohms
Low ESD Vinyl Cleanroom Chair
Tekniske data
Tekniske data
| Produkttype | Cleanroom ESD Chair |
| Farve | Black |
| Autoklaverbar | Not Autoclavable |
| Rygstil | Ergonomic |
| Stil | Castors |
| Med armlæn | No |
| Med ryglæn | Yes |
| Med hjul | Yes |
| Fodstøtte | No |
| Med lændestøtte | Yes |
| Vis mere |