missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ WNTinib
Description
WNTinib is a multi-kinase inhibitor with selectivity for mutant β-catenin (CTNNB1) in hepatocellular carcinoma compared to wild type hepatocytes (IC50 ranges are 0.36 μM-0.54 μM in primary organoids; 0.28 μM-1 μM in primary human HCC cell lines and 0.45 μM-2.3 μM in human cell lines ). Also inhibits VEGFR2, PDE6D and VEGFR1 (IC50 values are 0.3 μM, 0.3 μM and 0.6 μM respectively). Downregulates WNT and phospho-EZH2, upregulates trimethylation of H3K27 and normalises expression of apoptotic genes.
Specifications
Specifications
| Kemisk navn eller materiale | 4-[3-Fluoro-4-[[[[4-(1,1,2,2,2-pentafluoroethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide |
| CAS | 2770091-44-4 |
| Mængde | 10 mg |
| Mål | beta-Catenin |
| Molekylær formel | C22H16F6N4O3 |
| Renhed | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?