missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ WNT9A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103482
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Antibody detects endogenous levels of total WNT9A.
WNT9A is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. WNT9A is likely to signal over only few cell diameters.
Tekniske data
| WNT9A | |
| Polyclonal | |
| Unconjugated | |
| WNT9A | |
| MGC138165; MGC141991; Protein Wnt-14; Protein Wnt-9a; wingless related MMTV integration site 9A; wingless-type MMTV integration site 14; wingless-type MMTV integration site 9A; wingless-type MMTV integration site family member 9A; wingless-type MMTV integration site family, member 14; wingless-type MMTV integration site family, member 9A; Wnt family member 9A; WNT14; wnt-14; Wnt9a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 216795, 7483 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14904, Q8R5M2 | |
| WNT9A | |
| A synthesized peptide derived from human WNT9A(Accession O14904), corresponding to amino acid residues V290-Q340. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion