missing translation for 'onlineSavingsMsg'
Få mere at vide

Wnt-C59, TRC

Artikelnummer. 30453387
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30453387 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30453387 Leverandør Toronto Research Chemicals Leverandørnr. TRCW4993005MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 4-(2-Methyl-4-pyridinyl)-N-[4-(3-pyridinyl)phenyl]-benzeneacetamide
CAS 1243243-89-1
Molekylær formel C8 H11 N5 O3
Mængde 5 mg
Synonym Wnt-C59, 4-(2-Methyl-4-pyridinyl)-N-[4-(3-pyridinyl)phenyl]-benzeneacetamide
InChI formel InChI=1S/C8H11N5O3/c1-5-6(13(15)16)7(14)11-8(10-5)9-4-12(2)3/h4H,1-3H3,(H,10,11,14)/b9-4-
SMIL CN(C)\C=N/C1=NC(=C(C(=O)N1)[N+](=O)[O-])C
IUPAC navn N,N-dimethyl-N'-(4-methyl-5-nitro-6-oxo-1H-pyrimidin-2-yl)methanimidamide
Molekylvægt (g/mol) 225.205
Formel vægt 225.086
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.