missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress WKYMVM

Product Code. 16492028
Encompass_Preferred
Change view
Click to view available options
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16492028 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492028 Supplier MedChemExpress Supplier No. HYP1120

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WKYMVM is a potent N-formyl peptide receptor (FPR1) and FPRL1/2 agonist, also activates several leukocyte effector functions such as chemotaxis, mobilization of complement receptor-3, and activation of the NADPH oxidase.

Specifications

Renhed 99.79%
CAS 187986-17-0
Indhold og opbevaring Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Til brug med (applikation) COVID-19-immunoregulation
Molekylvægt (g/mol) 856.11 g/mol
Produkttype Peptides
Mængde 5 mg
Forsendelsestilstand Room temperature
Opløselighed H2O : 12.5 mg/mL (14.60 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.