Learn More
SP Wilmad-LabGlass™ High-Throughput NMR Tubes
Product Code. 16685672
Change view
Click to view available options
Quantity:
100 Pack
50 Pack
Unit Size:
Pack of 100
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16685672 100 Pack Pack of 100
16695672 50 Pack Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16685672 Supplier SP Supplier No. 669995702
  • ONLINE EXCLUSIVE PRICE
    1145.00 DKK
    1100.79 DKK / Pakke med 100
    Spare 44.21 DKK
    3% Slukket
Forventet forsendelse: 15-12-2026
Double the writing area allows for easy sample marking and identification.
  • Average camber of 60 microns ensures spectrum clarity for small molecule (MW <250) samples up to 600 MHz.
  • 2x larger marking area eliminates cramped and unreadable sample names
  • Silkscreened writing area keeps sample names from easily wiping off
  • Designed for routine use in most low to mid field NMR spectrometers
  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • Made from ASTM E438 Type 1 Class B glass
  • 100% inspected for surface defects and physical dimensions
Note: For experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.
Specifications
Length (Metric) 178 mm
Material Glass (Type 1, Class B)
Product Type High Throughput NMR Tube
Diameter (Metric) Outer 5±0.02 mm
Length (English) 7 in.
Thickness (Metric) Wall 0.43 mm
Quantity 100 Pack