Beskrivelse
- Breathable back panel to help improve air ventilation and reduce heat build-up.
- Knit cuffs provide a soft wrist closure for increased comfort compared to elastic closures.
- Elastic waist and ankles for convenience and freedom of movement.
- Two-way zipper with sealable storm flap for convenience and to help provide additional protection.
Tekniske data
Tekniske data
| Produkttype | General Purpose Coveralls |
| Størrelse | 3X-Large |
| Køn | Unisex |
| Engangs | Single-use |
| Talje stil | Elastic |
| Manchet stil | Knit |
| Hals stil | Hooded |
| Sterilitet | Non-sterile |
| Beskyttelsesklasse | Category III |
| Model- | 4535W |
| Vis mere |