Learn More
DWK Life Sciences Wheaton™ Straight Plug Style Silicone Stoppers for Vials and Serum Bottles
For maximum sealing of vials with straight wall glass finish
Brand: DWK Life Sciences 224100-178
5657.25 DKK valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 1.81440kg
Description
- Available in choice of different materials (silicone and bromobutyl)
Not recommended for vials with a blow-back feature.
Specifications
Straight Plug Stopper | |
Silicone | |
55 Durometer |
2cm | |
Fits 1.3cm I.D., 2cm O.D. mouth of vials and serum bottles | |
Yes |