Learn More
DWK Life Sciences Wheaton™ Straight Plug Rubber Stoppers
Provides maximum sealing for vials with straight-wall glass finishes. Wheaton™ Straight Plug Rubber Stoppers are manufactured from the highest quality raw materials.
Brand:  DWK Life Sciences 224100-177
Additional Details : Weight : 1.36080kg
 View more versions of this product
Product Code. 10476413
Quantity Price
1 3546.00 DKK / Pack of 100
6 3357.35 DKK / Pack of 100
Estimated Shipment
06-05-2021
Add to basket
Description
Description
  • Autoclavable
  • Available in a variety of sizes/colors
  • Manufactured to yield exceptional dimensional stability
Specifications
Specifications
Vacule Ampules
20 mm
Black
Straight Plug Stopper
Viton™