Learn More
DWK Life Sciences Wheaton™ Nephelo Culture Flask with Cleanout Port and Caps
Features depressed side arm and side clean-out port
Brand:  DWK Life Sciences 351514
1339.49 DKK valid until 2025-12-31
Use promo code "24329" to get your promotional price.
 View more versions of this product
Product Code. 12411381
  • 1490.00 DKK / Each
Only null left
Estimated Shipment: 10-11-2025
Description
  • Low-extractable borosilicate glass
  • Side arm size, 19 Ă— 140mm
  • Cap sizes, 38mm-430 (top cap) and 20mm-415 (side arm cap)
  • Caps have white rubber liner
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 mL
Turbidity meter or nephelometer
5.5 in.
430
38 mm
Borosilicate Glass
14 x 140 mm Sidearm
14 cm
Autoclavable
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Nephelo Culture Flask with Cleanout Port and Caps > Capacity: 1000mL; Sidearm Size: 14 x 140mm