Learn More
DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips (15mm)
15mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)
Brand: DWK Life Sciences W242427-A
This item is not returnable.
View return policy
Description
- Extended controlled dropper tip dispenses one drop at a time
- Average drop size is 44μL ±6μL
- Case of 1000
Specifications
Purple | |
1000 Cs. | |
41 ÎĽL | |
Plastic dropping bottle |
Polyethylene | |
ISO 9001 | |
Dropper Tip |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips (15mm) > Purple; 1000/Cs.
Spot an opportunity for improvement?Share a Content Correction