Learn More
Cytiva Whatman™ Glass Microfiber Filter Accessories
Filter Funnel Replacement Reservoirs for the 3-Piece Filter Funnel.
Brand: Cytiva 1950-207
1106.75 DKK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.11000kg
Specifications
Glass microfiber filter | |
Filter Funnel Reservoir | |
2.7 in. | |
7 cm |