Learn More
Cytiva Whatman™ Glass Fiber Filter Papers - Grade 31
Offers fine particle retention and high flow rate, as well as good loading capacity
Brand: Cytiva 10372803
359.10 DKK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.02000kg
Specifications
Glass Fiber | |
Fine particle retention and high flow rate, as well as good loading capacity | |
2.1 in. |
Fast | |
31 | |
5.5 cm |