Learn More
Cytiva Whatman™ Cellulose Nitrate Membrane Filters
Manufactured under strictly controlled conditions. Whatman™ Cellulose Nitrate Membrane Filters have a noticeably improved flexibility and are made to tolerate abuse during handling, loading, and autoclaving without sacrificing integrity.
Brand:  Cytiva 10402014
1558.00 DKK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.04000kg
 View more versions of this product
Product Code. 11385154
  • 1640.00 DKK / Pack of 100
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
Among the strongest of their type available, as measured and compared by burst pressure tests.
  • Narrow pore size distribution for improved surface capture and analysis.
  • Low levels of extractables to ensure sample integrity.
  • High strength and flexibility
  • Increased temperature stability
  • Reduced shrinkage
Specifications
Specifications
Disc
0.1 μm
105 to 140 μm
50 mm
Cellulose Nitrate
100 Pack
1.9 in.