Learn More
Cytiva Whatman™ Anodisc™ Filter Membranes
Constructed with precision pore structure and narrow pore size distribution. Whatman™ Anodisc™ Filter Membranes ensure a high level of particle removal efficiency. Captures microorganisms and particulates on the surface of the membrane for subsequent analysis.
3555.00 DKK - 6915.00 DKK
Product Code | Brand | Diameter (Metric) | Pore Size | Includes | Autoclavable | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Pore Size | Includes | Autoclavable | Price | Quantity & Availability | ||
11334734
|
Cytiva
6809-7023 |
13 mm | 0.2 μm | Without support ring | Autoclavable |
4340.00 DKK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
11324734
|
Cytiva
6809-7013 |
13 mm | 0.1 μm | Without support ring | Autoclavable |
4410.00 DKK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
11314734
|
Cytiva
6809-7003 |
13 mm | 0.02 μm | Without support ring | Autoclavable |
4750.00 DKK
Pack of 100 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
11335044
|
Cytiva
6809-6012 |
25 mm | 0.1 μm | Polypropylene support ring | Not Autoclavable |
3555.00 DKK
Pack of 50 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
10480713
|
Cytiva
6809-6002 |
25 mm | 0.02 μm | Polypropylene support ring | Not Autoclavable |
5130.00 DKK
Pack of 50 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
11304734
|
Cytiva
6809-6022 |
25 mm | 0.2 μm | Polypropylene support ring | Not Autoclavable |
4575.00 DKK
Pack of 50 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
11384724
|
Cytiva
6809-5022 |
47 mm | 0.2 μm | Polypropylene support ring | Not Autoclavable |
6505.00 DKK
Pack of 50 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
11374724
|
Cytiva
6809-5012 |
47 mm | 0.1 μm | Polypropylene support ring | Not Autoclavable |
6915.00 DKK
Pack of 50 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
11364724
|
Cytiva
6809-5002 |
47 mm | 0.02 μm | Polypropylene support ring | Not Autoclavable |
6695.00 DKK
Pack of 50 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
11954416
|
Cytiva
6809-5522 |
47 mm | 0.2 μm | Without support ring | Not Autoclavable |
5315.00 DKK
Pack of 50 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
11345044
|
Cytiva
6809-5502 |
47 mm | 0.02 μm | Without support ring | Not Autoclavable |
5360.00 DKK
Pack of 50 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||
Descripción
Constructed with precision pore structure and narrow pore size distribution. Whatman™ Anodisc™ Filter Membranes ensure a high level of particle removal efficiency. Captures microorganisms and particulates on the surface of the membrane for subsequent analysis.
Anopore inorganic membrane circles available with or without support ring contain no lateral crossovers between individual pores.- Wide solvent compatibility reduces the need to stock a variety of membranes in the laboratory.
- No additives used in the manufacturing process ensures minimal extractables and no sample contamination.
- Extremely low protein binding minimizes sample loss.
- Virtually transparent when wet making it highly suitable for microscopy studies.
Especificaciones
Aluminum Oxide |