Learn More
Welch™ Compact Aspiration/Filtration Vacuum Stations: Pipette, Receiver Kit and Filter
Accumulate waste liquids safe and easily. Aspiration systems prevent ingestion of fluids leading to vacuum pump failure.
Brand:  Welch™ U1475K-21
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 12759606
  • 2612.90 DKK / Pack of 10
Estimated Shipment: 02-10-2024
to see stock.
Add to basket
Includes: 40.5 oz. (1.2L) autoclaveable collection receiver, a hydrophobic inline filter, an automatic shutoff, vacuum regulator and gauge
Description
Description
  • Bleach resistant construction for extending service life
  • Continuous vacuum level monitoring with dial vacuum gauge
  • Powered by WOB-L piston vacuum pump
Recommended for aspirating aqueous and bleach solutions and not for acidic, basic or organic vapors and gases
Specifications
Specifications
Welch Compact Aspiration/Fitlration Vacuum Station
Replacement Hydrophobic Filter