Learn More
Welch™ Compact Aspiration/Filtration Vacuum Stations: Pipette, Receiver Kit and Filter
Accumulate waste liquids safe and easily. Aspiration systems prevent ingestion of fluids leading to vacuum pump failure.
Brand:  Welch™ U1475K-20
Additional Details : Weight : 30.00000kg
 View more versions of this product
Product Code. 10523543
  • 1865.00 DKK / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket
Includes: 40.5 oz. (1.2L) autoclaveable collection receiver, a hydrophobic inline filter, an automatic shutoff, vacuum regulator and gauge
Description
Description
  • Bleach resistant construction for extending service life
  • Continuous vacuum level monitoring with dial vacuum gauge
  • Powered by WOB-L piston vacuum pump
Recommended for aspirating aqueous and bleach solutions and not for acidic, basic or organic vapors and gases
Specifications
Specifications
6 ft. Tubing, 1.2L autoclavable jar with lid and two hydrophobic filters
Welch Compact Aspiration/Fitlration Vacuum Station
Spare Receiver Kit