Få mere at vide
Sampling Systems Weighing Boats
Shop alle Sampling Systems produkter
Kapacitet (metrisk):
25 ml
50 ml
Sterilitet:
Ikke-steril
Steril
Beskrivelse
- Made from high quality food grade PS
- Manufactured in a class 8 cleanroom, packed in a class 7 cleanroom
- Sterile and non-sterile options available
- Available from stock
- Fast delivery worldwide
Tekniske data
Tekniske data
| Kapacitet (engelsk) | 1.7 oz. |
| Autoklaverbar | Not Autoclavable |
| Bund form | Flat |
| Kapacitet (metrisk) | 50 ml |
| Farve | White |
| Engangs | Single-use |
| Materiale | Polystyrene |
| Form | Boat |
| Sterilitet | Ikke-steril |
| Type | Weighing Boat |