missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Wee1, GST Tag Recombinant Protein
WEE1 is classified as a Ser/Thr kinase by sequence analysis.
Brand: Invitrogen™ PR7373A
12445.00 DKK valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
WEE1 has been shown to inactivate multiple CDKs by tyrosine phosphorylation.
Specifications
NP_003381.1 | |
99.1 kDa | |
WEE1 | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: Other | |
WEE1 | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
100μg | |
WEE Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Baculovirus | |
Recombinant, Full Length | |
Signal Transduction |