missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ WDR45B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568659
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 93%.
This gene encodes a member of the WIPI or SVP1 family of WD40 repeat-containing proteins. The protein contains seven WD40 repeats that are thought to fold into a beta-propeller structure that mediates protein-protein interactions, and a conserved motif for interaction with phospholipids. The human genome contains several pseudogenes of this gene.
Tekniske data
| WDR45B | |
| Polyclonal | |
| Unconjugated | |
| WDR45B | |
| 0610008N23Rik; AA408735; D16Bwg0193e; RGD1305141; WD repeat domain 45B; WD repeat domain phosphoinositide-interacting protein 3; WD repeat protein 45-like; WD repeat-containing protein 45B; WD repeat-containing protein 45-like; Wdr45 like; WDR45B; Wdr45l; WDR45-like; WDR45-like protein; WIPI3; WIPI-3; WIPI49-like protein; zgc:56002 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56270 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5MNZ6 | |
| WDR45B | |
| synthetic peptide directed towards the middle region of human WDR45B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion