missing translation for 'onlineSavingsMsg'
Learn More

WBP2 Mouse anti-Human, CoraLite 488, Clone: 2B4G7, Proteintech

Mouse Monoclonal Antibody

1013.48 DKK - 2734.91 DKK

Specifications

Antigen WBP2
Clone 2B4G7
Applications Immunofluorescence
Classification Monoclonal
Conjugate CoraLite 488
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17791289
View Documents Promotion Details
Proteintech
CL488-66585-20UL
20 μL
1013.48 DKK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
17649866
View Documents Promotion Details
Proteintech
CL488-66585-100UL
100 μL
2734.91 DKK
100µL
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

WBP2 Monoclonal antibody apecifically detetects WBP2 antigen in Human Samples. It is validated for Immunofluorescence.
Specifications

Specifications

WBP2
Immunofluorescence
CoraLite 488
Mouse
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
23558
WBP2 fusion protein Ag2661
Primary
Store at -20°C. Avoid exposure to light.
2B4G7
Monoclonal
Liquid
Human
BC010616
WBP2
IgG1
Protein G purification
Videos
SDS
Documents

Documents

Product Certifications