Learn More
Sartorius Waymouth's MB 752/1 Medium
Product Code. 31908486 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. Quantity Unit Size
31908486 500 mL 500mL
1 options
Product Code. 31908486 Supplier Sartorius Supplier No. 011101A
 Restricted Item
På lager
Designed specifically for the cultivation of mouse L929 fibroblast cells in a chemically defined, serum-free synthetic medium.
  • Today's Waymouth's applicability is much more extensive and includes the establishment of carcinoma cell lines from pleural effusions and the growth of potentially tumorigenic cells.
  • It is also used for whole organ culture.
  • Modified as follows:
    • With Glucose
    • With Phenol Red
    • With L-Glutamine
    • With Sodium Bicarbonate
Specifications
Form Liquid
Product Type Cell Culture Media
Quantity 500 mL