Få mere at vide
Tocris Bioscience™ WAY 316606 hydrochloride
Secreted Frizzled-related protein-1 (sFRP-1) inhibitor; increases Wnt signaling
Brand: Tocris Bioscience™ 4767/10
Beskrivelse
WAY 316606 hydrochloride is a secreted Frizzled-Related Protein-1 (sFRP-1) inhibitor (IC50 = 0.65 μM); prevents sFRP-1 from interacting with Wnt and thus increases Wnt signaling. Increases total bone area in a murine calvarial organ culture assay.We are aware of a study in PLOS Biology where WAY-316606 was found to stimulate hair follicle growth. Please note that WAY-316606 from Tocris is for laboratory research use only, and that we will only sell to established scientific businesses and institutes.
Tekniske data
| >99% | |
| 1781835-02-6 | |
| 10 mg |
For Research Use Only