Få mere at vide
Tocris Bioscience™ WAY 267464 dihydrochloride
Beskrivelse
Potent, selective non-peptide oxytocin receptor (OTR) agonist (Ki = 58.4 nM at human OTR). Exhibits anxiolytic-like effects. May have therapeutic potential for the treatment of psychosis and schizophrenia.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | WAY 267464 dihydrochloride |
| CAS | 1432043-31-6 |
| Mængde | 10 mg |
| Molekylær formel | C32H37Cl2N7O4 |
| Formel vægt | 654.59 |
| Renhed | >97% |
| Molekylvægt (g/mol) | 654.593 |