Få mere at vide
Palintest™ Water Testing
Artikelnummer. 13159280
Format:
Tablet
Pakningsstørrelse:
Pakke med 250
Artikelnummer. 13159280
Brand: Palintest™ AS 096
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
These water testing tablets are ideal for quick and reliable testing in the field.
  • For calcium and magnesium ion results hardness expressed in ppm of calcium carbonate 
  • Ideal for quick and reliable testing in the field 
  • They are crushed in the water sample until a color change occurs and the number required allows results to be calculated in ppm or mg/L
  • Magnesium hardness is the difference between total and calcium hardness
  • Permanent hardness is the difference between total hardness and total alkalinity
  • In bottles of 250 tablets with instructions
Tekniske data
Format Tablet
Emballage Flaske
Mængde 250 stk