Learn More
Bochem™ Water Bath with Lid
Product Code. 10595221
Change view
Click to view available options
Capacity (Metric):
5.0 L
13.0 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10595221 5.0 L Each
10028330 13.0 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10595221 Supplier Bochem™ Supplier No. 8466
Forventet forsendelse: 10-12-2026
Pan and lid style water bath with two supporting handles. Bochem™ Water Bath with Lid is constructed of 18/10 stainless steel.
  • Pan and lid.
  • 18/10 stainless steel construction.
Specifications
Capacity (Metric) 5.0 L
Includes Lid
Material Stainless Steel