missing translation for 'onlineSavingsMsg'
Få mere at vide

Warangalone, MedChemExpress

Artikelnummer. 16482559
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16482559 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16482559 Leverandør Medchem Express Leverandørnr. HYN10741mg

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Warangalone is an anti-malarial compound which can inhibit the growth of both strains of parasite 3D7 (chloroquine sensitive) and K1 (chloroquine resistant) with IC50s of 4.8 μg/mL and 3.7 μg/mL, respectively. Warangalone can also inhibit cyclic AMP-dependent protein kinase catalytic subunit (cAK) with an IC50 of 3.5 μM.

Warangalone is an anti-malarial compound which can inhibit the growth of both strains of parasite 3D7 (chloroquine sensitive) and K1 (chloroquine resistant) with IC50s of 4.8 μg/mL and 3.7 μg/mL, respectively. Warangalone can also inhibit cyclic AMP-dependent protein kinase catalytic subunit (cAK) with an IC50 of 3.5 μM.

Tekniske data

Kemisk navn eller materiale Warangalone
Fysisk form Solid
CAS 4449-55-2
Farve Yellow
Mængde 1 mg
Renhedsgrad noter Research
Molekylær formel C25H24O5
Til brug med (applikation) COVID-19-immunoregulation
Synonym Scandenolone
SMIL O=C1C(C(O)=C(C=CC(C)(C)O2)C2=C3C/C=C(C)\C)=C3OC=C1C4=CC=C(O)C=C4
Molekylvægt (g/mol) 404.46
Formel vægt 404.46
Procent renhed 98.0%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.