Learn More
Köttermann Wall-mounted Cabinet
Create additional storage space in any laboratory with this Wall-mounted Cabinet.
2657.42 DKK - 3084.92 DKK
Specifications
Cabinet Type | Wall Mounted Cabinet |
---|---|
Door Type | Hinged |
Finish | Epoxy Coated |
Hinge Side | Right |
Locking Mechanism | Yes |
Product Code | Brand | Dimensions (D x W x H) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (D x W x H) | Price | Quantity & Availability | |||||
17332099
|
Köttermann
307-00053 |
366 x 600 x 480 mm |
2657.42 DKK
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
17342099
|
Köttermann
307-00058 |
366 x 600 x 630 mm |
2836.51 DKK
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
17352099
|
Köttermann
307-00063 |
366 x 600 x 780 mm |
3084.92 DKK
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
Description
- Unobtrusive storage space
- Features lockable door
- Constructed of durable epoxy coated steel
Specifications
Wall Mounted Cabinet | |
Epoxy Coated | |
Yes | |
None | |
White |
Hinged | |
Right | |
1 | |
1 | |
Steel |