Learn More
Uvex™ Waist Coat 7427-Graphite
The uvex suXXeed collection gets everything spot on: well thought-out functionality, sporty design, exciting color contrasts, unusual material combinations and a cut that fits like a glove. Can be worn in industrial and metal processing settings as well as for leisure.
650.00 DKK
Specifications
Closure Type Zipper
Product Type Cold-resistant Vest
Color Gray
Thickness (Metric) 245 g/m²
View More Specs
Products 8
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17227071
View Documents
Uvex™
8970908
X-Small
650.00 DKK
Each
Only null left
Estimated Shipment: 15-07-2025
Add to basket
17237071
View Documents
Uvex™
8970909
Small
650.00 DKK
Each
Only null left
Estimated Shipment: 15-07-2025
Add to basket
17247071
View Documents
Uvex™
8970910
Medium
650.00 DKK
Each
Only null left
Estimated Shipment: 15-07-2025
Add to basket
17257071
View Documents
Uvex™
8970911
Large
650.00 DKK
Each
Only null left
Estimated Shipment: 15-07-2025
Add to basket
17267071
View Documents
Uvex™
8970912
X-Large
650.00 DKK
Each
17277071
View Documents
Uvex™
8970913
2X-Large
650.00 DKK
Each
Only null left
Estimated Shipment: 15-07-2025
Add to basket
17287071
View Documents
Uvex™
8970914
3X-Large
650.00 DKK
Each
Only null left
Estimated Shipment: 15-07-2025
Add to basket
17297071
View Documents
Uvex™
8970915
4X-Large
650.00 DKK
Each
Only null left
Estimated Shipment: 15-07-2025
Add to basket
Description
Description
  • Waist Coat
  • Material: 97% polyester, 3% elasthan ca. 300 g/m²
  • The modern and sporty Soft Shell Waist Coat is characterized by maximum breathability and concurrently wind-resistance.
  • Ideal Waist Coat for intensive activities in cooler environs.
  • 4-way-stretch-material for an optimal fit and permanent well-being, pockets in the seams do offer enough storage space, extended back part for more safety in kidney areas, ideal as the second layer or as compensation for a jacket, suitable for industrial laundering
  • Ă–ko-Tex Standard 100
  • Sizes: 40/42 - 64/66
  • Care instructions: (state 19.09.2011) 30°C washable (gentle wash), chlorinate impossible, ironing impossible, dry cleaning impossible, tumble dryer: I point, suitable for industrial laundering
Specifications
Specifications
Zipper
Cold-resistant Vest
Gray
245 g/m²
Documents
Product Certifications
Product Content Correction
Product Title
Uvex™ Waist Coat 7427-Graphite