Få mere at vide
VX-765, TRC
Artikelnummer. 30456799
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456799 5 mg
30450147 50 mg
30453278 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30456799 Leverandør Toronto Research Chemicals Leverandørnr. TRCV9006005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 273404-37-8
Molekylær formel C24 H33 Cl N4 O6
Molekylvægt (g/mol) 509
Synonym L-Prolinamide, N-(4-amino-3-chlorobenzoyl)-3-methyl-L-valyl-N-[(2R,3S)-2-ethoxytetrahydro-5-oxo-3-furanyl]-, N-(4-Amino-3-chlorobenzoyl)-3-methyl-L-valyl-N-[(2R,3S)-2-ethoxytetrahydro-5-oxo-3-furanyl]-L-prolinamide, Belnacasan, VX 765
IUPAC navn (2S)-1-[(2S)-2-[(4-amino-3-chlorobenzoyl)amino]-3,3-dimethylbutanoyl]-N-[(2R,3S)-2-ethoxy-5-oxooxolan-3-yl]pyrrolidine-2-carboxamide
SMIL CCO[C@@H]1OC(=O)C[C@@H]1NC(=O)[C@@H]2CCCN2C(=O)[C@@H](NC(=O)c3ccc(N)c(Cl)c3)C(C)(C)C
Tekniske data
Kemisk navn eller materiale Belnacasan
Mængde 5 mg
InChI formel InChI=1S/C24H33ClN4O6/c1-5-34-23-16(12-18(30)35-23)27-21(32)17-7-6-10-29(17)22(33)19(24(2,3)4)28-20(31)13-8-9-15(26)14(25)11-13/h8-9,11,16-17,19,23H,5-7,10,12,26H2,1-4H3,(H,27,32)(H,28,31)/t16-,17-,19+,23+/m0/s1
Formel vægt 508.2089