Få mere at vide
Tocris Bioscience™ VU 6001966
Beskrivelse
VU 6001966 is a potent and selective mGlu2 negative allosteric modulator (IC50 = 78 nM). Exhibits > 350-fold selectivity for mGlu2 over mGlu3. Also exhibits selectivity over other mGluRs and a panel of GPCRs, ion channels and transporters. VU 6001966 induces antidepression like effects in mouse models of chronic stress. CNS penetrant.
Tekniske data
Tekniske data
| CAS | 2009052-76-8 |
| Mængde | 5 mg |
| Mål | Group II mGluR Modulators |
| Molekylær formel | C17H15FN4O2 |
| Formel vægt | 2424 |
| Renhed | 98% |